Merck
  • Pathogen-driven nucleotide overload triggers mitochondria-centered cell death in phagocytes.
Pathogen-driven nucleotide overload triggers mitochondria-centered cell death in phagocytes.
PLoS pathogens (2024-01-02)
Nicoletta Schwermann, Rita Haller, Sebastian Koch, Guntram A Grassl, Volker Winstel
ABSTRACT
Staphylococcus aureus is a dangerous pathogen that evolved refined immuno-evasive strategies to antagonize host immune responses. This involves the biogenesis of death-effector deoxyribonucleosides, which kill infectious foci-penetrating macrophages. However, the exact mechanisms whereby staphylococcal death-effector deoxyribonucleosides and coupled imbalances of intracellular deoxyribonucleotide species provoke immune cell death remain elusive. Here, we report that S. aureus systematically promotes an overload of deoxyribonucleotides to trigger mitochondrial rupture in macrophages, a fatal event that induces assembly of the caspase-9-processing apoptosome and subsequent activation of the intrinsic pathway of apoptosis. Remarkably, genetic disruption of this cascade not only helps macrophages coping with death-effector deoxyribonucleoside-mediated cytotoxicity but also enhances their infiltration into abscesses thereby ameliorating pathogen control and infectious disease outcomes in laboratory animals. Combined with the discovery of protective alleles in human CASP9, these data highlight the role of mitochondria-centered apoptosis during S. aureus infection and suggest that gene polymorphisms may shape human susceptibility toward a predominant pathogen.
Sigma-Aldrich
Anti-Staphylococcal Enterotoxin A antibody produced in rabbit, whole antiserum
Sigma-Aldrich
Anti-Staphylococcal α-Toxin antibody produced in rabbit, whole antiserum