Merck
  • Effect of xylitol tablets with and without red propolis on salivary parameters, dental biofilm and sensory acceptability of adolescents: a randomized crossover clinical trial.
Effect of xylitol tablets with and without red propolis on salivary parameters, dental biofilm and sensory acceptability of adolescents: a randomized crossover clinical trial.
Biofouling (2020-12-19)
Mariana Leonel Martins, Amanda Souza Nunes Monteiro, Thiago Isidro Vieira, Maria Bárbara de Carvalho Torres Guimarães, Letícia Coli Louvisse de Abreu, Lucio Mendes Cabral, Yuri Wanderley Cavalcanti, Lucianne Cople Maia, Andréa Fonseca-Gonçalves
ABSTRACT
The effect of xylitol tablets with (XPT) and without (XT) red propolis on salivary parameters, dental biofilm and acceptability of adolescents was evaluated through a blinded randomized crossover clinical trial. Healthy volunteers were allocated in the XPT and XT groups with a 30-day washout period for consumption of two tablets/dayfor seven days. An increase in salivary parameters was only observed immediately after tablet consumption, without differences between XPT and XT. The results for total microorganisms were similar, but XT was better in controlling Streptococcus spp. Rothia dentocariosa and Streptococcus salivarius were the most frequent in the biofilm and saliva, respectively. XPT and XT showed similar acceptability, with the highest purchase intention for XT. Although propolis did not enhance the properties of XT, further studies testing different protocols and follow-up are necessary; XT controlled Streptococcus spp. in biofilms, which demonstrate its potential for clinical application.
Product Description
Pyrex® Lifetime Red graduated tube, O.D. × L 25 mm × 200 mm, volume 50.0 mL, accuracy: 0.4 mL