Merck
  • BB-301: a silence and replace AAV-based vector for the treatment of oculopharyngeal muscular dystrophy.
BB-301: a silence and replace AAV-based vector for the treatment of oculopharyngeal muscular dystrophy.
Molecular therapy. Nucleic acids (2021-03-20)
Vanessa Strings-Ufombah, Alberto Malerba, Shih-Chu Kao, Sonal Harbaran, Fanny Roth, Ornella Cappellari, Ngoc Lu-Nguyen, Keiko Takahashi, Sophie Mukadam, Georgina Kilfoil, Claudia Kloth, Petrus Roelvink, George Dickson, Capucine Trollet, David Suhy
ABSTRACT
Oculopharyngeal muscular dystrophy (OPMD) is a rare autosomal dominant disease that results from an alanine expansion in the N-terminal domain of Poly-A Binding Protein Nuclear-1 (PABPN1). We have recently demonstrated that a two-vector gene therapy strategy significantly ameliorated the pathology in a mouse model of OPMD. This approach entailed intramuscular injection of two recombinant adeno-associated viruses (AAVs), one expressing three short hairpin RNAs (shRNAs) to silence both mutant and wild-type PABPN1 and one expressing a codon-optimized version of PABPN1 that is insensitive to RNA interference. Here we report the continued development of this therapeutic strategy by delivering "silence and replace" sequences in a single AAV vector named BB-301. This construct is composed of a modified AAV serotype 9 (AAV9) capsid that expresses a unique single bifunctional construct under the control of the muscle-specific Spc5-12 promoter for the co-expression of both the codon-optimized PABPN1 protein and two small inhibitory RNAs (siRNAs) against PABPN1 modeled into microRNA (miRNA) backbones. A single intramuscular injection of BB-301 results in robust inhibition of mutant PABPN1 and concomitant replacement of the codon-optimized PABPN1 protein. The treatment restores muscle strength and muscle weight to wild-type levels as well as improving other physiological hallmarks of the disease in a mouse model of OPMD.
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)−Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Laminin-2 (α-2 Chain) antibody, Rat monoclonal, clone 4H8-2, purified from hybridoma cell culture
Sigma-Aldrich
Anti-HSP90 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution