Merck
  • Periostin Promotes Colorectal Tumorigenesis through Integrin-FAK-Src Pathway-Mediated YAP/TAZ Activation.
Periostin Promotes Colorectal Tumorigenesis through Integrin-FAK-Src Pathway-Mediated YAP/TAZ Activation.
Cell reports (2020-01-23)
Handong Ma, Jing Wang, Xueli Zhao, Tiantian Wu, Zhengjie Huang, Dafan Chen, Yingfu Liu, Gaoliang Ouyang
ABSTRACT
Periostin is a multifunctional extracellular matrix protein involved in various inflammatory diseases and tumor metastasis; however, evidence regarding whether and how periostin actively contributes to inflammation-associated tumorigenesis remains elusive. Here, we demonstrate that periostin deficiency significantly inhibits the occurrence of colorectal cancer in azoxymethane/dextran sulfate sodium-treated mice and in ApcMin/+ mice. Moreover, periostin deficiency attenuates the severity of colitis and reduces the proliferation of tumor cells. Mechanistically, stromal fibroblast-derived periostin activates FAK-Src kinases through integrin-mediated outside-in signaling, which results in the activation of YAP/TAZ and, subsequently, IL-6 expression in tumor cells. Conversely, IL-6 induces periostin expression in fibroblasts by activating STAT3, which ultimately facilitates colorectal tumor development. These findings provide the evidence that periostin promotes colorectal tumorigenesis, and identify periostin- and IL-6-mediated tumor-stroma interaction as a promising target for treating colitis-associated colorectal cancer.
Sigma-Aldrich
Anti-Integrin αVβ3 Antibody, clone 27.1 (VNR-1), clone 27.1 (VNR-1), Chemicon®, from mouse
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Integrin αVβ5 Antibody, clone P1F6, azide free, clone P1F6, Chemicon®, from mouse
Pricing and availability is not currently available.