Merck
  • A liquid chromatography-tandem mass spectrometry (LC-MS/MS)-based assay to profile 20 plasma steroids in endocrine disorders.
A liquid chromatography-tandem mass spectrometry (LC-MS/MS)-based assay to profile 20 plasma steroids in endocrine disorders.
Clinical chemistry and laboratory medicine (2020-02-23)
Zhenxin Wang, Hao Wang, Yingfei Peng, Fangjun Chen, Lin Zhao, Xiaomu Li, Jiaqian Qin, Qianqian Li, Beili Wang, Baishen Pan, Wei Guo
ABSTRACT
Liquid chromatography-tandem mass spectrometry (LC-MS/MS)-based assays are employed in more and more clinical laboratories to quantify steroids. The steroid quantification by LC-MS/MS shows great value in screening or diagnosing endocrine disorders; however, the number of functional steroids included in the LC-MS/MS methods is still limited. Here, we describe the performance and validation of a 20-steroid plasma panel by LC-MS/MS. The panel included progestogens (including mineralocorticoids and glucocorticoids), androgens and estrogens biosynthesized in steroid metabolic pathways. The LC-MS/MS method was validated according to guidance documents, and subsequently employed to profile steroid changes in endocrine disorders. Using LC-MS/MS, 20 steroids were separated and quantified in 8 min. Coefficients of variation (CVs) of the 20 analytes at the lower limit of quantification (LLoQ) were all less than 15% (ranging from 1.84% to 14.96%). The linearity of the assay was demonstrated by all the R2 values greater than 0.995. Individual plasma steroids changed significantly in patients with subclinical Cushing's syndrome (SCS) and polycystic ovary syndrome (PCOS) - 17-hydroxypregnenolone (17-OH-PR), testosterone (T) and dihydrotestosterone (DHT) were significantly decreased in SCS patients, while in PCOS patients, pregnenolone, corticosterone (CORT), androstenedione (A4) and T were significantly increased and DHT was decreased. The LC-MS/MS method we developed for the quantification of 20 plasma steroids is clinical practicable. The steroid profiling data using this assay indicate its screening value for endocrine disorders. To further explore the value of the assay, more investigations are however needed.
Sigma-Aldrich
21-Deoxycortisol
Sigma-Aldrich
21-Hydroxyprogesterone, ≥97% (HPLC)
Sigma-Aldrich
Dehydroepiandrosterone-2,2,3,4,4,6-d6, 97 atom % D, 98% (CP)
Sigma-Aldrich
Reichstein′s substance S, ≥98%