Merck
  • Humanized mouse model of Rasmussen's encephalitis supports the immune-mediated hypothesis.
Humanized mouse model of Rasmussen's encephalitis supports the immune-mediated hypothesis.
The Journal of clinical investigation (2018-04-10)
Hania Kebir, Lionel Carmant, François Fontaine, Kathie Béland, Ciprian M Bosoi, Nathalie T Sanon, Jorge I Alvarez, Sébastien Desgent, Camille L Pittet, David Hébert, Marie-Josée Langlois, Rose-Marie Rébillard, Dang K Nguyen, Cécile Cieuta-Walti, Gregory L Holmes, Howard P Goodkin, John R Mytinger, Mary B Connolly, Alexandre Prat, Elie Haddad
ABSTRACT
Rasmussen's encephalitis (RE) is a chronic inflammatory brain disorder that causes frequent seizures and unilateral hemispheric atrophy with progressive neurological deficits. Hemispherectomy remains the only treatment that leads to seizure freedom for this refractory epileptic syndrome. The absence of an animal model of disease has been a major obstacle hampering the development of effective therapies. Here, we describe an experimental mouse model that shares several clinical and pathological features with the human disease. Immunodeficient mice injected with peripheral blood mononuclear cells from RE patients and monitored by video electroencephalography developed severe seizures of cortical origin and showed intense astrogliosis and accumulation of human IFN-γ- and granzyme B-expressing T lymphocytes in the brain compared with mice injected with immune cells from control subjects. We also provide evidence for the efficacy of α4 integrin blockade, an approved therapy for the treatment of multiple sclerosis and Crohn's disease, in reducing inflammatory markers associated with RE in the CNS. This model holds promise as a valuable tool for understanding the pathology of RE and for developing patient-tailored experimental therapeutics.
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein (GFAP)−Cy3 antibody, Mouse monoclonal, clone G-A-5, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Amyloid Precursor Protein, N-Terminal antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution