Skip to Content
Merck
trans-3-(3-Pyridyl)acrylic acid 99%

trans-3-(3-Pyridyl)acrylic acid

trans-3-(3-Pyridyl)acrylic acid 99%
Synonym(s):
3-Pyridineacrylic acid
Empirical Formula (Hill Notation):
C8H7NO2
CAS Number:
Molecular Weight:
149.15

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing