
Tiamulin hydrogen fumarate

Synonym(s):
Tiamulin fumarate, Thiamutilin fumarate
Empirical Formula (Hill Notation):
C32H51NO8S
CAS Number:
Molecular Weight:
609.81
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing