
Thiamphenicol

Synonym(s):
D-threo-2,2-Dichloro-N-(β-hydroxy-α-[hydroxymethyl]-4-[methylsulfonyl]phenethyl)acetamide
Empirical Formula (Hill Notation):
C12H15Cl2NO5S
CAS Number:
Molecular Weight:
356.22
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Purity
Activity Spectrum
Format
Quality Segment
Application
Mode of Action
Optical Isomer
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing