
PbS core-type quantum dots

Synonym(s):
CANdot®, Fluorescent nanocrystals, QDs, artificial atoms
Empirical Formula (Hill Notation):
PbS
Molecular Weight:
239.27
Products
Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Showing 1-4 of 4
Compare
Product Number
Description
Pricing