
NovaSeptum® GO High Purity Bag Sampling System, Manifold (5-valve)

Synonym(s):
NovaSeptum® GO High Purity Bag
Products
Product Category
Brand
Product Line
Agency
Quality Segment
Application
Sterility
Sterilization Method
Showing 1-1 of 1
Compare
Product Number
Description
Pricing