
Frovatriptan succinate

Synonym(s):
Frovatriptan succinate monohydrate, Frovatriptan succinate, R-(+) 3-Methylamino-6-carboxamido-1,2,3,4-tetrahydrocarbazole monosuccinate monohydrate
Empirical Formula (Hill Notation):
C14H17N3O · C4H6O4 · H2O
CAS Number:
Molecular Weight:
379.41
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing