
Fentanyl solution

Empirical Formula (Hill Notation):
C22H28N2O
CAS Number:
Molecular Weight:
336.47
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2