Skip to Content
Merck
Ethynyltrimethylsilane 98%

Ethynyltrimethylsilane

Ethynyltrimethylsilane 98%
Synonym(s):
Trimethylsilylacetylene
Linear Formula:
(CH3)3SiC≡CH
CAS Number:
Molecular Weight:
98.22

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing