Skip to Content
Merck
Ethylenediaminetetraacetic acid tetrasodium salt dihydrate purum, ≥98.0% (KT)

Ethylenediaminetetraacetic acid tetrasodium salt dihydrate

Ethylenediaminetetraacetic acid tetrasodium salt dihydrate purum, ≥98.0% (KT)
Synonym(s):
EDTA tetrasodium salt, Edathamil, Tetrasodium ethylenediaminetetraacetate dihydrate
Linear Formula:
(NaOOCCH2)2NCH2CH2N(CH2COONa)2 · 2H2O
CAS Number:
Molecular Weight:
416.20
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Reagent Type
Special Grade
Technique