
Dantrolene sodium

Synonym(s):
Dantrolene
sodium salt hydrate sodium salt hemi(heptahydrate), 1-[(5(p-Nitrophenyl)furfurylidene)amino]hydantoin sodium salt hemi(heptahydrate)
sodium salt hydrate sodium salt hemi(heptahydrate), 1-[(5(p-Nitrophenyl)furfurylidene)amino]hydantoin sodium salt hemi(heptahydrate)
Empirical Formula (Hill Notation):
C14H9N4NaO5 · 3.5H2O
CAS Number:
Molecular Weight:
399.29
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade