
Clofazimine for system suitability

Synonym(s):
Clofazimine, N,5-Bis(4-chlorophenyl)-3,5-dihydro-3-(isopropylimino)phenazin-2-amine
Empirical Formula (Hill Notation):
C27H22Cl2N4
CAS Number:
Molecular Weight:
473.40
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing