Skip to Content
Merck
Adenosine 5′-monophosphate monohydrate from yeast, ≥97%

Adenosine 5′-monophosphate monohydrate

Adenosine 5′-monophosphate monohydrate from yeast, ≥97%
Synonym(s):
5′-AMP, A-5′-P, AMP
Empirical Formula (Hill Notation):
C10H14N5O7P · H2O
CAS Number:
Molecular Weight:
365.24

Products

Brand
Biological Source
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing