
3-Morpholinosydnonimine hydrochloride

Synonym(s):
3-(4-Morpholinyl)sydnone imine hydrochloride, Linsidomine hydrochloride, SIN-1 hydrochloride
Empirical Formula (Hill Notation):
C6H10N4O2 · HCl
CAS Number:
Molecular Weight:
206.63
Products
Product Category
Brand
Formula Weight
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing