Skip to Content
Merck
2-Pentanol analytical standard

2-Pentanol

2-Pentanol analytical standard
Synonym(s):
(±)-2-Pentanol, sec-Pentyl alcohol, Methyl propyl carbinol, 2-Pentanol
Linear Formula:
CH3CH2CH2CH(OH)CH3
CAS Number:
Molecular Weight:
88.15
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Showing 1-4 of 4
Compare
Product Number
Description
Pricing