
2,6-Dimethylpyridine

Synonym(s):
2,6-Lutidine, 2,6-Dimethylpyridine
Linear Formula:
2,6-(CH3)2C5H3N
CAS Number:
Molecular Weight:
107.15
Products
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing