Skip to Content
Merck
All Photos(1)

Key Documents

CRM37038

Supelco

Flumorph

certified reference material, TraceCERT®, mixture of stereoisomers, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

(EZ)-4-[3-(3,4-Dimethoxyphenyl)-3-(4-fluorophenyl)acryloyl]morpholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H22FNO4
CAS Number:
Molecular Weight:
371.40
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

application(s)

environmental

format

neat

storage temp.

2-8°C

SMILES string

COc1ccc(cc1OC)\C(=C/C(=O)N2CCOCC2)c3ccc(F)cc3

InChI

1S/C21H22FNO4/c1-25-19-8-5-16(13-20(19)26-2)18(15-3-6-17(22)7-4-15)14-21(24)23-9-11-27-12-10-23/h3-8,13-14H,9-12H2,1-2H3/b18-14+

InChI key

BKBSMMUEEAWFRX-NBVRZTHBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.Download your certificate at: http://www.sigma-aldrich.com

Application

Flumorph is a systemic fungicide used mainly to control Oomycetes in a variety of crops as vines, vegetables and potatoes. It is not listed in the European Pesticide Database and not approved for its use as a plant protection product in the European Union (EU). The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service