Skip to Content
Merck
All Photos(1)

Key Documents

SAB1300027

Sigma-Aldrich

Anti-ANPA (N-term) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-ANP-A, Anti-ANPRA, Anti-Atrial natriuretic peptide receptor A, Anti-GC-A, Anti-Guanylatcyclase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, mouse, rat

technique(s)

indirect ELISA: 1:1000
indirect immunofluorescence: 1:10-1:50
western blot: 1:100-1:500

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ANPA(4881)

General description

ANPA is a receptor for atrial natriuretic peptide. It exhibits guanylate cyclase activity on binding of ANF. There seem to be at least three ANP receptors: two with guanylate cyclase activity (ANPA and ANPB) and one (ANPC) which is probably responsible for the clearance of ANP from the circulation without a role in signal transduction. This Type I membrane protein belongs to the adenylyl cyclase class-4/guanylyl cyclase family and contains 1 protein kinase-like domain.

Immunogen

ANPA (NP_000897, 1-30)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the N-terminal region of human ANPA.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service