Skip to Content
Merck
All Photos(1)

Key Documents

8C9

99072805, Reactivity: Anti-CD36, human

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

Mouse x Mouse Hybridoma

ImmunoDon

BALB/c mouse spleen,female

Myeloma

NS-1

Reactivity

Anti-CD36, human

technique(s)

cell culture | mammalian: suitable

isotype

IgM

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse x Mouse hybridoma

Cell Line Description

The hybridoma line 8C9 was produced by immunizing a female BALB/c mouse i.p. with tissue from a human malignant fibrous histiocytoma once a week for 6 weeks. Mouse spleen cells were fused with myeloma NS-1 cells using PEG and HAT for selection . The 8C9 monoclonal antibody detects a unique epitope on the leukocyte differentiation antigen CD36 (platelet glycoprotein IV or III6) expressed by normal and neoplastic endothelial cells. Immunoreactivity with 8C9 was detected on capillary endothelial cells, adipocytes, monocytes, platelets and the human monocytoid cell line U-937 (Sigma Catalogue number. 85011440). The antibody did not react with erythroid cells, T cells, B cells or HUVECs (human umbilical vein endothelial cells). Immunoblot analysis showed that 8C9 bound to a 97-kDa membrane protein expressed by U-937 cells treated with phorbol ester. Comparative immunohistochemical studies using 8C9 and the monoclonal antibody OKM demonstrated epitopic heterogeneity of the CD36 antigen by normal and neoplastic endothelial cells.

Immunogen

Human malignant fibrous histiocytoma

Application

Study of endothelial cells, CD36 expression analysis

Culture Medium

RPMI 1640 + 2mM glutamine + 10% FBS

Subculture Routine

Maintain cultures between 3-9 x 100,000 cells/ml; 5% CO2; 37°C

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service