Skip to Content
Merck
All Photos(1)

Key Documents

567280

Sigma-Aldrich

4-(3-Hydroxypropyl)morpholine

95%

Synonym(s):

3-(4-Morpholinyl)-1-propanol, 3-Morpholinopropanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H15NO2
CAS Number:
Molecular Weight:
145.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

95%

impurities

may contain ~5% morpholine

refractive index

n20/D 1.4770 (lit.)

bp

240-241 °C (lit.)

density

1.049 g/mL at 25 °C (lit.)

SMILES string

OCCCN1CCOCC1

InChI

1S/C7H15NO2/c9-5-1-2-8-3-6-10-7-4-8/h9H,1-7H2

InChI key

VZKSLWJLGAGPIU-UHFFFAOYSA-N

Application

4-(3-Hydroxypropyl)morpholine may be used in the synthesis of macrocyclic urea compounds, which can act as potent Chk1 inhibitors.

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Repr. 2 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

185.0 °F - closed cup

Flash Point(C)

85 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David Johansson et al.
BMC cancer, 9, 67-67 (2009-02-28)
The prerequisite for the potential use of the bacterial toxin verotoxin-1 in the treatment of breast cancer was investigated by first determining the expression of its receptor Gb3 (CD77) in clinical breast cancer tissue specimens. We then examined the cytotoxicity
"Synthesis and in-vitro biological activity of macrocyclic urea Chk1 inhibitors"
Li J, et al.
Bioorganic & Medicinal Chemistry Letters, 17(23), 6499-6504 (2007)
Y Jiang et al.
Journal of biological regulators and homeostatic agents, 34(6), 2029-2036 (2020-12-12)
Dysregulation of the adipo-osteogenic differentiation balance of mesenchymal stem cells (MSCs), which are common progenitor cells of adipocytes and osteoblasts, has been associated with many pathophysiologic diseases, such as obesity, osteopenia, and several neurodegenerative disorders. Growing evidence suggests that lipid
Weihong Xu et al.
Biochemical and biophysical research communications, 463(4), 545-550 (2015-06-01)
In the current study, we investigated the potential activity of AR-42, a novel histone deacetylase (HDAC) inhibitor, against colon cancer cells. Our in vitro results showed that AR-42 induced ceramide production, exerted potent anti-proliferative and pro-apoptotic activities in established (SW-620 and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service