GC Analysis of a 17-Component Dioxin Congener Mix on SLB®-5ms (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
SLB-5ms, 15 m x 0.10 mm I.D., 0.10 μm (28466-U)
oven
150 °C (1 min), 35 °C/min to 340 °C (1 min)
inj. temp.
250 °C
detector
ECD, 340 °C
carrier gas
hydrogen, 45 cm/sec, constant
injection
1 μL splitless (1 min)
liner
4 mm I.D., single taper
sample
17-component 2,3,7,8-substituted dioxin standard, 100-500 ppb in n-nonane
Description
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany