Merck
  • Vitamin D Receptor Regulates Amyloid Beta 1-42 Production with Protein Disulfide Isomerase A3.
Vitamin D Receptor Regulates Amyloid Beta 1-42 Production with Protein Disulfide Isomerase A3.
ACS chemical neuroscience (2017-07-15)
Duygu Gezen-Ak, Irem L Atasoy, Esin Candaş, Merve Alaylioglu, Selma Yılmazer, Erdinç Dursun
ABSTRACT
The challenge of understanding the biology of neuronal amyloid processing could provide a basis for understanding the amyloid pathology in Alzheimer's disease (AD). Based on our previous studies, we have suggested that AD might be the consequence of a hormonal imbalance in which the critical hormone is vitamin D. The present study primarily focused on the creation of a condition that prevents the genomic or nongenomic action of vitamin D by disrupting vitamin D receptors (VDR or PDIA3/1,25MARRS); the effects of these disruptions on the series of proteins involved in secretases that play a crucial role in amyloid pathology and on amyloid beta (Aβ) production in primary cortical neurons were observed. VDR and PDIA3/1,25MARRS genes were silenced separately or simultaneously in E16 primary rat cortical neurons. The expression of target genes involved in APP processing, including Presenilin1, Presenilin2, Nicastrin, BACE1, ADAM10, and APP, was investigated with qRT-PCR and Western blot in this model. 1,25-Dihydroxyvitamin D
Sigma-Aldrich
MISSION® esiRNA, targeting human PPIB
Sigma-Aldrich
Mechlorethamine hydrochloride, 98%