Merck
  • Anticancer copper pyridine benzimidazole complexes: ROS generation, biomolecule interactions, and cytotoxicity.
Anticancer copper pyridine benzimidazole complexes: ROS generation, biomolecule interactions, and cytotoxicity.
Journal of inorganic biochemistry (2016-12-05)
Kathleen E Prosser, Stephanie W Chang, Felix Saraci, Phuc H Le, Charles J Walsby
ABSTRACT
The Cu(II) complex CuCl
Sigma-Aldrich
4-Fluorobenzyl bromide, 97%
Sigma-Aldrich
Ethidium homodimer, suitable for fluorescence, ~90% (HPCE)
Sigma-Aldrich
Benzyl bromide, reagent grade, 98%