Merck
  • Role of pannexin 1 in Clostridium perfringens beta-toxin-caused cell death.
Role of pannexin 1 in Clostridium perfringens beta-toxin-caused cell death.
Biochimica et biophysica acta (2016-11-05)
Soshi Seike, Masaya Takehara, Keiko Kobayashi, Masahiro Nagahama
ABSTRACT
Beta-toxin produced by Clostridium perfringens is a key virulence factor of fatal hemorrhagic enterocolitis and enterotoxemia. This toxin belongs to a family of β-pore-forming toxins (PFTs). We reported recently that the ATP-gated P2X We examined the effect of Panx1 in beta-toxin-induced cell death utilizing selective antagonists, knockdown of Panx1, and binding using dot-blot analysis. Localization of Panx1 and the P2X Selective Panx1 antagonists (carbenoxolone [CBX], probenecid, and Panx1 inhibitory peptide) prevented beta-toxin-caused cell death in THP-1 cells. CBX did not block the binding of the toxin to cells. Small interfering knockdown of Panx1 blocked beta-toxin-mediated cell death through inhibiting the oligomer formation of the toxin. Beta-toxin triggered a transient ATP release from THP-1 cells, but this early ATP release was blocked by CBX. ATP scavengers (apyrase and hexokinase) inhibited beta-toxin-induced cytotoxicity. Furthermore, co-administration of ATP with beta-toxin enhanced the binding and cytotoxicity of the toxin. Based on our results, Panx1 activation is achieved through the interaction of beta-toxin with the P2X Pannexin 1 is a novel candidate therapeutic target for beta-toxin-mediated disease.
Sigma-Aldrich
10Panx1 trifluoroacetate salt, ≥98% (HPLC)
Sigma-Aldrich
5(6)-Carboxyfluorescein diacetate, BioReagent, suitable for fluorescence, ≥90.0% (HPLC)