Skip to Content
Beginning October 1st, 2024, the Life Science business of Merck Ltd. Taiwan will implement list price increases on our Science and Lab Solutions products. Learn More
Merck
  • Rhein Inhibits AlkB Repair Enzymes and Sensitizes Cells to Methylated DNA Damage.
Rhein Inhibits AlkB Repair Enzymes and Sensitizes Cells to Methylated DNA Damage.
The Journal of biological chemistry (2016-03-27)
Qi Li, Yue Huang, Xichun Liu, Jianhua Gan, Hao Chen, Cai-Guang Yang
ABSTRACT
The AlkB repair enzymes, including Escherichia coli AlkB and two human homologues, ALKBH2 and ALKBH3, are iron(II)- and 2-oxoglutarate-dependent dioxygenases that efficiently repair N(1)-methyladenine and N(3)-methylcytosine methylated DNA damages. The development of small molecule inhibitors of these enzymes has seen less success. Here we have characterized a previously discovered natural product rhein and tested its ability to inhibit AlkB repair enzymes in vitro and to sensitize cells to methyl methane sulfonate that mainly produces N(1)-methyladenine and N(3)-methylcytosine lesions. Our investigation of the mechanism of rhein inhibition reveals that rhein binds to AlkB repair enzymes in vitro and promotes thermal stability in vivo In addition, we have determined a new structural complex of rhein bound to AlkB, which shows that rhein binds to a different part of the active site in AlkB than it binds to in fat mass and obesity-associated protein (FTO). With the support of these observations, we put forth the hypothesis that AlkB repair enzymes would be effective pharmacological targets for cancer treatment.
Sigma-Aldrich
Anti-ALKBH2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Pricing and availability is not currently available.