Merck
  • Retrograde fluxes of focal adhesion proteins in response to cell migration and mechanical signals.
Retrograde fluxes of focal adhesion proteins in response to cell migration and mechanical signals.
Molecular biology of the cell (2007-09-07)
Wei-hui Guo, Yu-li Wang
ABSTRACT
Recent studies suggest that mechanical signals mediated by the extracellular matrix play an essential role in various physiological and pathological processes; yet, how cells respond to mechanical stimuli remains elusive. Using live cell fluorescence imaging, we found that actin filaments, in association with a number of focal adhesion proteins, including zyxin and vasodilator-stimulated phosphoprotein, undergo retrograde fluxes at focal adhesions in the lamella region. This flux is inversely related to cell migration, such that it is amplified in fibroblasts immobilized on micropatterned islands. In addition, the flux is regulated by mechanical signals, including stretching forces applied to flexible substrates and substrate stiffness. Conditions favoring the flux share the common feature of causing large retrograde displacements of the interior actin cytoskeleton relative to the substrate anchorage site, which may function as a switch translating mechanical input into chemical signals, such as tyrosine phosphorylation. In turn, the stimulation of actin flux at focal adhesions may function as part of a feedback mechanism, regulating structural assembly and force production in relation to cell migration and mechanical load. The retrograde transport of associated focal adhesion proteins may play additional roles in delivering signals from focal adhesions to the interior of the cell.
Sigma-Aldrich
Y-27632-CAS 331752-47-7-Calbiochem, Y-27632A, CAS 331752-47-7, is a cell-permeable, reversible, inhibitor of Rho kinases (Ki = 140 nM for p160ROCK). Enhances survival & cloning efficiency of ESC without affecting their pluripotency.
Sigma-Aldrich
Genistein, Soybean, A cell-permeable, reversible, substrate competitive inhibitor of protein tyrosine kinases, including autophosphorylation of epidermal growth factor receptor kinase (IC₅₀ = 2.6 µM).
Sigma-Aldrich
PP2, PP2, CAS 172889-27-9, is a potent, reversible, ATP-competitive, inhibitor of the Src family of protein tyrosine kinases (IC₅₀ = 4, 5, 5, &100 nM for p56lck, p59fynT, Hck, & Src, respectively).