Merck
[Ala1,3,11,15]endothelin-1 analogs with ETB agonistic activity.
Biochemical and biophysical research communications (1991-08-30)
T Saeki, M Ihara, T Fukuroda, M Yamagiwa, M Yano
ABSTRACT
A linear peptide analog of endothelin (ET)-1, [Ala1,3,11,15]ET-1 (4AlaET-1), and its truncated peptide analogs were synthesized to study the structural requirements of ET-1 for the recognition of ETs-nonselective ETB receptors. ET-1 exhibited sub-nanomolar binding to two distinct ET receptor subtypes (ETA and ETB), but 4AlaET-1 bound to ETB with an affinity 1,700 times higher than that seen during binding to ETA. The truncated linear peptides 4AlaET-1(6-21), 4AlaET-1(8-21) and N-acetyl-4AlaET-1(10-21) still had high affinity for ETB, whereas 4AlaET-1(6-20) and 4AlaET-1(11-21) displayed remarkably reduced affinity for ETB. Therefore, ET-1 requires the Glu10-Trp21 sequence for ETB binding, but not the disulfide bridges. These ETB-binding peptides elicit endothelium-dependent vasorelaxation of porcine pulmonary arteries in parallel with the binding affinity for ETB, suggesting that they are ETB agonists.