Merck
Cicatricial entropion associated with chronic dipivefrin application.
Ophthalmic plastic and reconstructive surgery (2001-10-20)
A O D'Ostroph, R A Dailey
ABSTRACT
To report patients who presented to the oculoplastics department for repair of cicatrical entropion after topical use of dipivefrin. To discuss the possible mechanisms of action and highlight the potential detrimental effects of dipivefrin on palpebral conjunctiva. Retrospective consecutive noncomparative case series. Nine eyes from 6 patients, 74 years to 90 years of age, referred by ophthalmologists for repair of cicatricial entropion after at least 2 years of twice-a-day application of dipivefrin. After cessation of topical dipivefrin application and successful surgical repair of entropion, no recurrence of signs or symptoms has been reported. Moderate lymphocytic infiltration of the substantia propria of the conjunctiva of both upper and lower lid specimens was present, as was scarring and keratinization of the epithelium. Cicatrization in the substantia propria of the conjunctiva by excessive lymphocytic infiltration after topically administered antiglaucoma drugs including dipivefrin is a possible mechanism of action for entropion.
Product Description
Dipivefrine for system suitability, European Pharmacopoeia (EP) Reference Standard
Dipivefrine hydrochloride, European Pharmacopoeia (EP) Reference Standard