Merck
  • Autoxidation of 4-methylcatechol: a model for the study of the biosynthesis of copper amine oxidases quinonoid cofactor.
Autoxidation of 4-methylcatechol: a model for the study of the biosynthesis of copper amine oxidases quinonoid cofactor.
Biochemical and biophysical research communications (1995-09-14)
A C Rinaldi, M C Porcu, N Curreli, A Rescigno, A Finazzi-Agró, J Z Pedersen, A Rinaldi, E Sanjust
ABSTRACT
The autoxidation of 4-methylcatechol under quasi-physiological conditions, leading to 2-hydroxy-5-methyl-1,4-benzoquinone, was investigated. The effects of pH and metal ions were examined. An electrophilic attack of dioxygen to the 4-methylcatechol monoanion to form a transient peroxo species is proposed. It was concluded that such a non-enzymic conversion is likely for this model compound and for its physiological counterpart, a specific tyrosyl residue incorporated in the protein chain at the active site of copper amine oxidases.
Product Description
USP
Levadopa Related Compound A, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
6-Hydroxy-DL-DOPA, ≥98% (HPLC), powder