Merck
  • Quantitative probing of surface charges at dielectric-electrolyte interfaces.
Quantitative probing of surface charges at dielectric-electrolyte interfaces.
Lab on a chip (2013-02-16)
Weihua Guan, Nitin K Rajan, Xuexin Duan, Mark A Reed
ABSTRACT
The intrinsic charging status at the dielectric-electrolyte interface (DEI) plays a critical role for electrofluidic gating in microfluidics and nanofluidics, which offers opportunities for integration of wet ionics with dry electronics. A convenient approach to quantitatively probe the surface charges at the DEI for material pre-selection purpose has been lacking so far. We report here a low-cost, off-chip extended gate field effect transistor configuration for direct electrostatic probing the charging status at the DEI. Capacitive coupling between the surface charges and the floating extended gate is utilized for signal transducing. The relationship between the surface charge density and the experimentally accessible quantities is given by device modeling. The multiplexing ability makes measuring a local instead of a globally averaged surface charge possible.
Sigma-Aldrich
Silicon nitride, predominantly β-phase, ≤10micron primary particle size
Sigma-Aldrich
Silicon nitride, predominantly α-phase, ≤10 micron
Sigma-Aldrich
Silicon nitride, nanopowder, <50 nm particle size (spherical), ≥98.5% trace metals basis
Sigma-Aldrich
Silicon nitride, powder, ≥99.9% trace metals basis