Merck
  • Sequential versus triple therapy for the first-line treatment of Helicobacter pylori: a multicentre, open-label, randomised trial.
Sequential versus triple therapy for the first-line treatment of Helicobacter pylori: a multicentre, open-label, randomised trial.
Lancet (London, England) (2012-11-20)
Jyh-Ming Liou, Chieh-Chang Chen, Mei-Jyh Chen, Chien-Chuan Chen, Chi-Yang Chang, Yu-Jen Fang, Ji-Yuh Lee, Shih-Jer Hsu, Jiing-Chyuan Luo, Wen-Hsiung Chang, Yao-Chun Hsu, Cheng-Hao Tseng, Ping-Huei Tseng, Hsiu-Po Wang, Ueng-Cheng Yang, Chia-Tung Shun, Jaw-Town Lin, Yi-Chia Lee, Ming-Shiang Wu
ABSTRACT
Whether sequential treatment can replace triple therapy as the standard treatment for Helicobacter pylori infection is unknown. We compared the efficacy of sequential treatment for 10 days and 14 days with triple therapy for 14 days in first-line treatment. For this multicentre, open-label, randomised trial, we recruited patients (≥20 years of age) with H pylori infection from six centres in Taiwan. Using a computer-generated randomisation sequence, we randomly allocated patients (1:1:1; block sizes of six) to either sequential treatment (lansoprazole 30 mg and amoxicillin 1 g for the first 7 days, followed by lansoprazole 30 mg, clarithromycin 500 mg, and metronidazole 500 mg for another 7 days; with all drugs given twice daily) for either 10 days (S-10) or 14 days (S-14), of 14 days of triple therapy (T-14; lansoprazole 30 mg, amoxicillin 1 g, and clarithromycin 500 mg for 14 days; with all drugs given twice daily). Investigators were masked to treatment allocation. Our primary outcome was the eradication rate in first-line treatment by intention-to-treat (ITT) and per-protocol (PP) analyses. This trial is registered with ClinicalTrials.gov, number NCT01042184. Between Dec 28, 2009, and Sept 24, 2011, we enrolled 900 patients: 300 to each group. The eradication rate was 90·7% (95% CI 87·4-94·0; 272 of 300 patients) in the S-14 group, 87·0% (83·2-90·8; 261 of 300 patients) in the S-10 group, and 82·3% (78·0-86·6; 247 of 300 patients) in the T-14 group. Treatment efficacy was better in the S-14 group than it was in the T-14 group in both the ITT analysis (number needed to treat of 12·0 [95% CI 7·2-34·5]; p=0·003) and PP analyses (13·7 [8·3-40], p=0·003). We recorded no significant difference in the occurrence of adverse effects or in compliance between the three groups. Our findings lend support to the use of sequential treatment as the standard first-line treatment for H pylori infection. National Taiwan University Hospital and National Science Council.
Product Description
Lansoprazole for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Lansoprazole, ≥98% (TLC), powder
Lansoprazole, European Pharmacopoeia (EP) Reference Standard