Merck
  • Membrane transport mechanisms of quinidine and procainamide in renal LLC-PK1 and intestinal LS180 cells.
Membrane transport mechanisms of quinidine and procainamide in renal LLC-PK1 and intestinal LS180 cells.
Biological & pharmaceutical bulletin (2010-08-06)
Miki Masago, Mari Takaai, Jumpei Sakata, Asuka Horie, Toshikazu Ito, Kazuya Ishida, Masato Taguchi, Yukiya Hashimoto
ABSTRACT
The aim of the present study was to compare the membrane transport mechanisms of procainamide with those of quinidine using renal epithelial LLC-PK(1) and intestinal epithelial LS180 cells. In LLC-PK(1) cells, the transcellular transport of 10 microM quinidine in the basolateral-to-apical direction was similar to that in the opposite direction, and 1 mM tetraethylammonium (TEA) did not affect the transcellular transport of the drug. On the other hand, the transcellular transport of 10 microM TEA and procainamide in LLC-PK(1) cells was directional from the basolateral side to the apical side. In addition, this directional transcellular transport of procainamide was diminished in the presence of 1 mM TEA. In LS180 cells, the temperature-dependent cellular uptake of 100 microM quinidine and procainamide was markedly increased by alkalization of the apical medium, and was inhibited significantly by 1 mM several hydrophobic cationic drugs, but not by TEA. The rank order of the inhibitory effects of hydrophobic cationic drugs on the uptake of procainamide in LS180 cells was imipramine>quinidine>diphenhydramine asymptotically equal topyrilamine>procainamide, which was consistent with that on the uptake of quinidine. These findings suggested that procainamide (but not quinidine) was transported by cation transport systems in renal epithelial cells, but that both procainamide and quinidine were taken up by another cation transport system in intestinal epithelial cells.
Product Description
Supelco
Procainamide hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material