Merck
  • High quality x-ray absorption spectroscopy measurements with long energy range at high pressure using diamond anvil cell.
High quality x-ray absorption spectroscopy measurements with long energy range at high pressure using diamond anvil cell.
The Review of scientific instruments (2009-08-07)
Xinguo Hong, Matthew Newville, Vitali B Prakapenka, Mark L Rivers, Stephen R Sutton
ABSTRACT
We describe an approach for acquiring high quality x-ray absorption fine structure (XAFS) spectroscopy spectra with wide energy range at high pressure using diamond anvil cell (DAC). Overcoming the serious interference of diamond Bragg peaks is essential for combining XAFS and DAC techniques in high pressure research, yet an effective method to obtain accurate XAFS spectrum free from DAC induced glitches has been lacking. It was found that these glitches, whose energy positions are very sensitive to the relative orientation between DAC and incident x-ray beam, can be effectively eliminated using an iterative algorithm based on repeated measurements over a small angular range of DAC orientation, e.g., within +/-3 degrees relative to the x-ray beam direction. Demonstration XAFS spectra are reported for rutile-type GeO2 recorded by traditional ambient pressure and high pressure DAC methods, showing similar quality at 440 eV above the absorption edge. Accurate XAFS spectra of GeO2 glass were obtained at high pressure up to 53 GPa, providing important insight into the structural polymorphism of GeO2 glass at high pressure. This method is expected be applicable for in situ XAFS measurements using a diamond anvil cell up to ultrahigh pressures.
Sigma-Aldrich
Germanium(IV) oxide, ≥99.99% trace metals basis
Sigma-Aldrich
Germanium(IV) oxide, powder, 99.999% trace metals basis
Sigma-Aldrich
Germanium(IV) oxide, (crystalline powder), 99.998% trace metals basis