Merck
  • Soluble precursors of 2,3-naphthalocyanine and phthalocyanine for use in thin film transistors.
Soluble precursors of 2,3-naphthalocyanine and phthalocyanine for use in thin film transistors.
Chemical communications (Cambridge, England) (2008-10-03)
Atsuko Hirao, Taiji Akiyama, Tetsuo Okujima, Hiroko Yamada, Hidemitsu Uno, Yoshimasa Sakai, Shinji Aramaki, Noboru Ono
ABSTRACT
Soluble precursors of 2,3-naphthalocyanine (Nc) and phthalocyanine (Pc) were prepared and were converted into insoluble semiconducting thin films of Pc and Nc by heating after fabrication via spin-coating.
Sigma-Aldrich
2,3-Naphthalocyanine, Dye content 95 %