Merck
  • Fluoxetine reduces L-DOPA-derived extracellular DA in the 6-OHDA-lesioned rat striatum.
Fluoxetine reduces L-DOPA-derived extracellular DA in the 6-OHDA-lesioned rat striatum.
Neuroreport (2001-05-08)
H Yamato, K Kannari, H Shen, T Suda, M Matsunaga
ABSTRACT
We investigated the effect of fluoxetine, a selective serotonin reuptake inhibitor (SSRI), on L-DOPA-derived extracellular dopamine (DA) levels in the striatum of rats with nigrostriatal dopaminergic denervation using in vivo microdialysis. Treatment with fluoxetine (10 mg/kg, i.p.) induced a 41% reduction in the cumulative amount of extracellular DA during 300 min following L-DOPA administration (50 mg/kg, i.p.; p < 0.01). This effect was antagonized by pretreatment with WAY-100635, a potent 5-HT1A antagonist, indicating that this effect of fluoxetine is due to its indirect 5-HT1A agonistic property. These results suggest that SSRIs may impair motor functions in patients with Parkinson's disease by reducing efflux of exogenous L-DOPA-derived DA.
Product Description
USP
Levadopa Related Compound A, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
6-Hydroxy-DL-DOPA, ≥98% (HPLC), powder