Merck
  • Mdm2 requires Sprouty4 to regulate focal adhesion formation and metastasis independent of p53.
Mdm2 requires Sprouty4 to regulate focal adhesion formation and metastasis independent of p53.
Nature communications (2024-08-21)
Rafaela Muniz de Queiroz, Gizem Efe, Asja Guzman, Naoko Hashimoto, Yusuke Kawashima, Tomoaki Tanaka, Anil K Rustgi, Carol Prives
ABSTRACT
Although the E3 ligase Mdm2 and its homologue and binding partner MdmX are the major regulators of the p53 tumor suppressor protein, it is now evident that Mdm2 and MdmX have multiple functions that do not involve p53. As one example, it is known that Mdm2 can regulate cell migration, although mechanistic insight into this function is still lacking. Here we show in cells lacking p53 expression that knockdown of Mdm2 or MdmX, as well as pharmacological inhibition of the Mdm2/MdmX complex, not only reduces cell migration and invasion, but also impairs cell spreading and focal adhesion formation. In addition, Mdm2 knockdown decreases metastasis in vivo. Interestingly, Mdm2 downregulates the expression of Sprouty4, which is required for the Mdm2 mediated effects on cell migration, focal adhesion formation and metastasis. Further, our findings indicate that Mdm2 dampening of Sprouty4 is a prerequisite for maintaining RhoA levels in the cancer cells that we have studied. Taken together we describe a molecular mechanism whereby the Mdm2/MdmX complex through Sprouty4 regulates cellular processes leading to increase metastatic capability independently of p53.
Sigma-Aldrich
Anti-phospho-Focal Adhesion Kinase (Tyr397) Antibody, clone 18, clone 18, from mouse
Sigma-Aldrich
5,6-Dichlorobenzimidazole 1-β-D-ribofuranoside
Sigma-Aldrich
MDM2 Inhibitor VII, MEL23, The MDM2 Inhibitor VII controls the biological activity of MDM2. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.