Merck
  • 4,4-Dimethyl- and diastereomeric 4-hydroxy-4-methyl- (2S)-glutamate analogues display distinct pharmacological profiles at ionotropic glutamate receptors and excitatory amino acid transporters.
4,4-Dimethyl- and diastereomeric 4-hydroxy-4-methyl- (2S)-glutamate analogues display distinct pharmacological profiles at ionotropic glutamate receptors and excitatory amino acid transporters.
ChemMedChem (2009-09-05)
Lennart Bunch, Darryl S Pickering, Thierry Gefflaut, Virginie Vinatier, Virgil Helaine, Ahmad Amir, Birgitte Nielsen, Anders A Jensen
ABSTRACT
Subtype-selective ligands are of great interest to the scientific community, as they provide a tool for investigating the function of one receptor or transporter subtype when functioning in its native environment. Several 4-substituted (S)-glutamate (Glu) analogues were synthesized, and altogether this approach has provided important insight into the structure-activity relationships (SAR) for ionotropic and metabotropic glutamate receptors (iGluRs and mGluRs), as well as the excitatory amino acid transporters (EAATs). In this work, three 4,4-disubstituted Glu analogues 1-3, which are hybrid structures of important 4-substituted Glu analogues 4-8, were investigated at iGluRs and EAATs. Collectively, their pharmacological profiles add new and valuable information to the SAR for the iGluRs and EAAT1-3.
Sigma-Aldrich
(4R)-4-Hydroxy-L-glutamic acid, ≥98.0% (TLC)
Pricing and availability is not currently available.