Merck
  • Gene expression profiles in granuloma tissue reveal novel diagnostic markers in sarcoidosis.
Gene expression profiles in granuloma tissue reveal novel diagnostic markers in sarcoidosis.
Experimental and molecular pathology (2014-04-29)
George P Christophi, Tiffany Caza, Christopher Curtiss, Divya Gumber, Paul T Massa, Steve K Landas
ABSTRACT
Sarcoidosis is an immune-mediated multisystem disease characterized by the formation of non-caseating granulomas. The pathogenesis of sarcoidosis is unclear, with proposed infectious or environmental antigens triggering an aberrant immune response in susceptible hosts. Multiple pro-inflammatory signaling pathways have been implicated in mediating macrophage activation and granuloma formation in sarcoidosis, including IFN-γ/STAT-1, IL-6/STAT-3, and NF-κB. It is difficult to distinguish sarcoidosis from other granulomatous diseases or assess disease severity and treatment response with histopathology alone. Therefore, development of improved diagnostic tools is imperative. Herein, we describe an efficient and reliable technique to classify granulomatous disease through selected gene expression and identify novel genes and cytokine pathways contributing to the pathogenesis of sarcoidosis. We quantified the expression of twenty selected mRNAs extracted from formalin-fixed paraffin embedded (FFPE) tissue (n = 38) of normal lung, suture granulomas, sarcoid granulomas, and fungal granulomas. Utilizing quantitative real-time RT-PCR we analyzed the expression of several genes, including IL-6, COX-2, MCP-1, IFN-γ, T-bet, IRF-1, Nox2, IL-33, and eotaxin-1 and revealed differential regulation between suture, sarcoidosis, and fungal granulomas. This is the first study demonstrating that quantification of target gene expression in FFPE tissue biopsies is a potentially effective diagnostic and research tool in sarcoidosis.