Merck
  • CKLF1/CCR5 axis is involved in neutrophils migration of rats with transient cerebral ischemia.
CKLF1/CCR5 axis is involved in neutrophils migration of rats with transient cerebral ischemia.
International immunopharmacology (2020-05-24)
Chen Chen, Shi-Feng Chu, Qi-di Ai, Zhao Zhang, Nai-Hong Chen
ABSTRACT
Chemokine-like factor 1 (CKLF1) is a chemokine increased significantly in ischemic brain poststroke. It shows chemotaxis effects on various immune cells, but the mechanisms of CKLF1 migrating neutrophils are poorly understood. Recent studies have provided evidence that CC chemokine receptor 5 (CCR5), a receptor of CKLF1, is involved in ischemic stroke. To investigate the effects of HIF-1α guided AAV in ischemic brain, investigating the outcome of stroke, and examining the involvement of CKLF1/CCR5 axis in recruitment of neutrophils. HIF-1α guided AAV knocked down CKLF1 in ischemic area and alleviated brain damage of rats. CKLF1 migrated neutrophils through CCR5, worsening inflammatory responses. Akt/GSK-3β pathway may involve in CKLF1/CCR5 axis guided neutrophils chemotaxis. CKLF1/CCR5 axis is involved in neutrophils migration of rats with transient cerebral ischemia. CKLF1/CCR5 axis may be a useful target for stroke therapy.
Sigma-Aldrich
3,7-Diacetyl-1,3,7-triaza-5-phosphabicyclo[3.3.1]nonane, 97%
Sigma-Aldrich
Akt1/2 kinase inhibitor, ≥98% (HPLC)
Sigma-Aldrich
TWS119, ≥98% (HPLC)