Merck
  • Cochaperone Mzb1 is a key effector of Blimp1 in plasma cell differentiation and β1-integrin function.
Cochaperone Mzb1 is a key effector of Blimp1 in plasma cell differentiation and β1-integrin function.
Proceedings of the National Academy of Sciences of the United States of America (2018-09-28)
Virginia Andreani, Senthilkumar Ramamoorthy, Abhinav Pandey, Ekaterina Lupar, Stephen L Nutt, Tim Lämmermann, Rudolf Grosschedl
ABSTRACT
Plasma cell differentiation involves coordinated changes in gene expression and functional properties of B cells. Here, we study the role of Mzb1, a Grp94 cochaperone that is expressed in marginal zone (MZ) B cells and during the terminal differentiation of B cells to antibody-secreting cells. By analyzing Mzb1 -/- Prdm1 +/gfp mice, we find that Mzb1 is specifically required for the differentiation and function of antibody-secreting cells in a T cell-independent immune response. We find that Mzb1-deficiency mimics, in part, the phenotype of Blimp1 deficiency, including the impaired secretion of IgM and the deregulation of Blimp1 target genes. In addition, we find that Mzb1 -/- plasmablasts show a reduced activation of β1-integrin, which contributes to the impaired plasmablast differentiation and migration of antibody-secreting cells to the bone marrow. Thus, Mzb1 function is required for multiple aspects of plasma cell differentiation.
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution