Merck
  • Challenges in 3D culturing of neutrophils: Assessment of cell viability.
Challenges in 3D culturing of neutrophils: Assessment of cell viability.
Journal of immunological methods (2018-02-25)
Miriam Gonzalez Gonzalez, Iwona Cichon, Anna Scislowska-Czarnecka, Elzbieta Kolaczkowska
ABSTRACT
Standard cell culturing on plastic plates (two dimensional (2D) cultures) does not represent the actual microenvironment where cells reside in tissues. The three dimensional (3D) systems, composed of extracellular matrix and/or pure amino acids which form a scaffold for cells, are more accurate in this respect. 3D cultures were primarily developed for cancer cells but there is also a need for their application in studies on inflammatory leukocytes. Herein we describe our approach to study neutrophil-like cells in the 3D system. We describe measures taken to establish a neutrophil-like cell line (nHL-60) and selection of 3D scaffolds (PuraMatrix alone or enriched with collagen type I) for their culturing. We focus on challenges in measurement of neutrophil viability in 3D cultures and based on our data we suggest application of resazurin, rather than tetrazolium-based dyes or trypan blue exclusion, for evaluation of neutrophil viability.
Sigma-Aldrich
L-Glutamine–Penicillin–Streptomycin solution, L-glutamine 200 mM, streptomycin 10 mg/mL, penicillin 10,000 units, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology