Skip to Content
Merck
p-Toluenesulfonamide Pharmaceutical Secondary Standard; Certified Reference Material

p-Toluenesulfonamide

p-Toluenesulfonamide Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
p-Toluenesulfonamide
Linear Formula:
CH3C6H4SO2NH2
CAS Number:
Molecular Weight:
171.22
EC Number:

Products

Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Technique