
Cyclizine impurity B

Synonym(s):
Diphenylmethanol, Benzhydrol, Benzhydryl alcohol, Diphenyl carbinol
Linear Formula:
(C6H5)2CHOH
CAS Number:
Molecular Weight:
184.23
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing