Crotonaldehyde-2,4-DNPH
Synonym(s):
Crotonaldehyde-2,4-dinitrophenylhydrazone
Linear Formula:
CH3CH=CHCH=NNHC6H3(NO2)2
CAS Number:
Molecular Weight:
250.21
Products
Brand
Formula Weight
Manufacturer
Melting Point (Ā°C)
Format
Quality Segment
Package Size
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing