
Carbonylimidazole, polymer-bound

Synonym(s):
4-Benzyloxybenzyl imidazole-1-carboxylate polymer-bound, Activated carbonate polymer-bound, Imidazole carbonate, polymer-bound, Imidazole carbonate, polymer-bound to Wang resin, Imidazole carboxylate-Wang resin, Wang imidazolide carbamate resin
Products
Brand
Quality Segment
Cleavage Condition
Linker
Chain Type
Reaction Type
Reactivity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing